Learn More
Description
Specifications
Specifications
| Antigen | TLR7 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF, KnockDown |
| Classification | Monoclonal |
| Clone | 4G6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 10ug/ml∼, Flow Cytometry 1-2ug/10^6cells, Immunohistochemistry 1:50 - 1:200. Use reported in scientific literature (PMID 31092820), Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Flow (Intracellular) 5 ug/ml, CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 31730654) |
| Gene Accession No. | Q9NYk1 |
| Gene Alias | toll-like receptor 7 |
| Gene Symbols | TLR7 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
