Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody (4G6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP227332SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227332SS 0.025 mg
NBP227332 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227332SS Supplier Novus Biologicals Supplier No. NBP227332SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), CyTOF-ready, Knockdown Validated.

Specifications

Antigen TLR7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF, KnockDown
Classification Monoclonal
Clone 4G6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 10ug/ml∼, Flow Cytometry 1-2ug/10^6cells, Immunohistochemistry 1:50 - 1:200. Use reported in scientific literature (PMID 31092820), Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Flow (Intracellular) 5 ug/ml, CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 31730654)
Gene Accession No. Q9NYk1
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Mouse
Immunogen A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51284
Test Specificity Human. Mouse reactivity reported in scientific literature (PMID: 24126361).
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.