Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody (4G6), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP225274V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225274V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225274V Supplier Novus Biologicals Supplier No. NBP225274V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen TLR7
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone 4G6
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, KnockDown Validated
Gene Accession No. Q9NYk1
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Mouse
Immunogen A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51284
Test Specificity Human. Mouse reactivity reported in scientific literature (PMID: 24126361).
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.