Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224906SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224906SS 0.025 mg
NBP224906 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224906SS Supplier Novus Biologicals Supplier No. NBP224906SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 67 publications

TLR7 Polyclonal specifically detects TLR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dot Blot, Flow (Intracellular), Bioactivity.

Specifications

Antigen TLR7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, Dot Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 2-5 ug/ 1x10^6 cells, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 28219705), Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation reported in scientific literature (PMID 17452530), Immunohistochemistry-Paraffin 1:200. Use reported in scientific literature (PMID 25264223), Immunohistochemistry-Frozen 1:10-1:2000. Use reported by customer review, Proximity Ligation Assay reported in scientific literature (PMID 30579042), Dot Blot reported in scientific literature (PMID 27248820), Flow (Intracellular), Bioactivity reported in scientific literature (PMID 28894449)
Formulation PBS with 0.02% Sodium Azide
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 706-749 of human TLR7; GenBank no. gb AAF78035.1 AF245702_1. It will cross-react with mouse TLR7. In human Ramos cells, additional bands are seen.
Purification Method Immunogen affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51284
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.