Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 67 publications

$257.00 - $596.50

Specifications

Antigen TLR7
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 2-5 ug/ 1x10^6 cells, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 28219705), Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation reported in scientific literature (PMID 17452530), Immunohistochemistry-Paraffin 1:200. Use reported in scientific literature (PMID 25264223), Immunohistochemistry-Frozen 1:10-1:2000. Use reported by customer review, Proximity Ligation Assay reported in scientific literature (PMID 30579042), Dot Blot reported in scientific literature (PMID 27248820), Flow (Intracellular), Bioactivity reported in scientific literature (PMID 28894449)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, Dot Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224906SS
SDP
View Documents
Novus Biologicals
NBP224906SS
0.025 mg
Each for $257.00
Only null left
Add to Cart
 
NBP224906
SDP
View Documents
Novus Biologicals
NBP224906
0.1 mg
Each for $596.50
Only null left
Add to Cart
 
Description

Description

TLR7 Polyclonal specifically detects TLR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dot Blot, Flow (Intracellular), Bioactivity.
Specifications

Specifications

TLR7
Western Blot 1-3 ug/ml, Flow Cytometry 2-5 ug/ 1x10^6 cells, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 28219705), Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation reported in scientific literature (PMID 17452530), Immunohistochemistry-Paraffin 1:200. Use reported in scientific literature (PMID 25264223), Immunohistochemistry-Frozen 1:10-1:2000. Use reported by customer review, Proximity Ligation Assay reported in scientific literature (PMID 30579042), Dot Blot reported in scientific literature (PMID 27248820), Flow (Intracellular), Bioactivity reported in scientific literature (PMID 28894449)
Polyclonal
Purified
RUO
Human, Mouse, Rat
toll-like receptor 7
TLR7
IgG
Immunogen affinity purified
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, Dot Blot
Unconjugated
Rabbit
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
PBS with 0.02% Sodium Azide
51284
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 706-749 of human TLR7; GenBank no. gb AAF78035.1 AF245702_1. It will cross-react with mouse TLR7. In human Ramos cells, additional bands are seen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.