Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224767SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224767SS 0.025 mg
NBP224767 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224767SS Supplier Novus Biologicals Supplier No. NBP224767SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

TLR7 Polyclonal specifically detects TLR7 in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TLR7
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Flow Cytometry 1-2 ug/ 10^6 cells, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25586463)
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Rabbit
Immunogen A portion of amino acids 900-950 of human TLR7 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51284
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and rhesus monkey, 83% homologous in dog and 72% homologous in mouse.
Target Species Human, Primate, Rhesus Macaque
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.