Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR8 Antibody (44C143), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP224917Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224917Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224917Y Supplier Novus Biologicals Supplier No. NBP224917AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR8 Monoclonal specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen TLR8
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 44C143
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Flow (Intracellular)
Gene Accession No. Q9NR97
Gene Alias CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
Gene Symbols TLR8
Host Species Mouse
Immunogen This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51311
Test Specificity This antibody is reactive to Human, Mouse.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.