Learn More
Description
Specifications
Specifications
| Antigen | TLR8 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, CyTOF |
| Classification | Monoclonal |
| Clone | 44C143 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, Simple Western 20 ug/ml, Flow Cytometry 0.5-1 ug/10^6 cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml, Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in multiple pieces of scientific literature, Flow (Intracellular) reported in reported in multiple pieces of scientific literature (PMID 24836676), CyTOF-ready |
| Gene Accession No. | Q9NR97 |
| Gene Alias | CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 |
| Gene Symbols | TLR8 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
