Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR8 Antibody (44C143) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 45 publications

$257.00 - $524.00

Specifications

Antigen TLR8
Clone 44C143
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Simple Western 20 ug/ml, Flow Cytometry 0.5-1 ug/10^6 cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml, Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in multiple pieces of scientific literature, Flow (Intracellular) reported in reported in multiple pieces of scientific literature (PMID 24836676), CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224917SS
SDP
View Documents
Novus Biologicals
NBP224917SS
0.025 mg
Each for $257.00
Only null left
Add to Cart
 
NBP224917
SDP
View Documents
Novus Biologicals
NBP224917
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
Description

Description

TLR8 Monoclonal specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready.
Specifications

Specifications

TLR8
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, CyTOF
Unconjugated
Mouse
Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors
Q9NR97
51311
This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
44C143
Western Blot 1-3 ug/ml, Simple Western 20 ug/ml, Flow Cytometry 0.5-1 ug/10^6 cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml, Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in multiple pieces of scientific literature, Flow (Intracellular) reported in reported in multiple pieces of scientific literature (PMID 24836676), CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse, Rabbit
CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
TLR8
IgG1 κ
Protein G purified
This antibody is reactive to Human, Mouse.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.