Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR8 Antibody (935166), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB302505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302505 Supplier Novus Biologicals Supplier No. FAB8999R

Mouse Monoclonal Antibody

TLR8 Monoclonal antibody specifically detects TLR8 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen TLR8
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 935166
Conjugate Alexa Fluor 647
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human TLR8, Glu27-Thr827, Accession # Q9NR97
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51311
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.