Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Antibody (1138D), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB373865 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373865 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373865 Supplier Novus Biologicals Supplier No. FAB7960I
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TLR9 Monoclonal antibody specifically detects TLR9 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry

Specifications

Antigen TLR9
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 1138D
Conjugate Janelia Fluor 549
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TLR9, Leu26-Asp818, Accession # AAK29625
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 54106
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.