Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Antibody (26C593.2), Biotin, Novus Biologicals™
SDP

Catalog No. NB337318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337318 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337318 Supplier Novus Biologicals Supplier No. NBP224729B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR9 Monoclonal antibody specifically detects TLR9 in Human, Mouse, Rat, Canine, Equine, Monkey, Primate samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay, In vitro Assay, Dot Blot, Flow (Intracellular), CyTOF, KnockDown, Neutralization

Specifications

Antigen TLR9
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 26C593.2
Conjugate Biotin
Dilution Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional, In vitro assay, Dot Blot, Flow (Intracellular), CyTOF-ready, Knockdown Validated, Block/Neutralize
Formulation PBS
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Host Species Mouse
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 54106
Target Species Human, Mouse, Rat, Canine, Equine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.