Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Antibody (26C593.2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224729 0.1 mg
NBP224729SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224729 Supplier Novus Biologicals Supplier No. NBP224729
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 137 publications

TLR9 Monoclonal specifically detects TLR9 in Canine, Equine, Human, Monkey, Mouse, Pig, Primate, Rat samples. It is validated for Block/Neutralize, CyTOF, Cytometric Bead Assay Standard, Dot Blot, ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Flow Cytometry Control, Functional Assay, IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown, Simple Western, Western Blot.

Specifications

Antigen TLR9
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay, In vitro Assay, Dot Blot, CyTOF, KnockDown, Neutralization
Classification Monoclonal
Clone 26C593.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml, Simple Western 30 ug/ml, Flow Cytometry, ELISA 1:100 - 1:2000. Use reported in multiple pieces of scientific literature, Immunohistochemistry reported in scientific literature (PMID 27744078), Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 25871979), Immunohistochemistry-Paraffin 5 ug/ml, Functional reported in scientific literature (PMID 25411258), In vitro assay reported in scientific literature (PMID 25411258), Dot Blot reported in scientific literature (PMID 27248820), Flow (Intracellular) 1:10 - 1:1000. Use reported in scientific literature (PMID 24986635), CyTOF-ready, KnockDown Validated reported in scientific literature (PMID 31655343), Block/Neutralize reported in scientific literature (PMID 25338738)
Gene Accession No. Q9NR96
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Gene Symbols TLR9
Host Species Mouse
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 54106
Test Specificity This antibody is reactive to Dog, Horse, Human, Mouse, Rat and Rhesus Monkey.
Target Species Canine, Equine, Human, Monkey, Mouse, Pig, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.