Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Antibody (26C593R), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB371744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371744 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371744 Supplier Novus Biologicals Supplier No. FAB36583E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR9 Monoclonal antibody specifically detects TLR9 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen TLR9
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 26C593R
Conjugate DyLight 405
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Host Species Mouse
Immunogen Antibody developed against the KLH-conjugated synthetic peptide corresponding to amino acids Asp250-Asn300 of human TLR9 isoform A., Accession # Q9NR96
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 54106
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.