Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Antibody (2A4C6.2E5), PerCP, Novus Biologicals™
SDP

Catalog No. NB337079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337079 Supplier Novus Biologicals Supplier No. NBP231150PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TLR9 Monoclonal antibody specifically detects TLR9 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ICC/IF (Negative)

Specifications

Antigen TLR9
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 2A4C6.2E5
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative)
Formulation PBS
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Host Species Rat
Immunogen Partial recombinant protein made to an internal portion of mouse TLR9 (between amino acids 500-800) [UniProt Q9EQU3]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 54106
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.