Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Antibody (5G5), Biotin, Novus Biologicals™
SDP

Catalog No. NB6001420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001420 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001420 Supplier Novus Biologicals Supplier No. NB6001420

Mouse Monoclonal Antibody

TLR9 Monoclonal antibody specifically detects TLR9 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoassay.

Specifications

Antigen TLR9
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone 5G5
Concentration 0.1 mg/mL
Conjugate Biotin
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay
Formulation PBS and 0.1% BSA with 0.02% Sodium Azide
Gene Accession No. Q9NR96
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Gene Symbols TLR9
Host Species Mouse
Immunogen Purified fusion protein of extracellular domain of human TLR9 (AA 1-815) and human IgGFc
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 54106
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.