Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLX/NR2E1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TLX/NR2E1 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TLX/NR2E1 Polyclonal specifically detects TLX/NR2E1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TLX/NR2E1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS buffer, 2% sucrose | |
7101 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
Polyclonal | |
Purified | |
RUO | |
Human | |
hTll, nuclear receptor subfamily 2, group E, member 1, Nuclear receptor TLX, Protein tailless homolog, tailes-related receptor, tailless homolog, Tll, TLXnuclear receptor subfamily 2 group E member 1, XTLL | |
The immunogen is a synthetic peptide directed towards the N terminal region of human TLX/NR2E1 (NP_003260). Peptide sequence MSKPAGSTSRILDIPCKVCGDRSSGKHYGVYACDGCSGFFKRSIRRNRTY | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title