Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLX2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB183633 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183633 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183633 Supplier Novus Biologicals Supplier No. NBP282023JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TLX2 Polyclonal antibody specifically detects TLX2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TLX2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias homeo box 11-like 1, Homeobox protein Hox-11L1, HOX11L1Tlx2, NCXEnx, Neural crest homeobox protein, T-cell leukemia homeobox 2, T-cell leukemia homeobox protein 2, T-cell leukemia, homeobox 2
Host Species Rabbit
Immunogen TLX2 antibody was raised against an 18 amino acid peptide near the amino terminus of human TLX2. The immunogen is located within amino acids 20 - 70 of TLX2. Amino Acid Squence: LGRGGQGHGENGAFSGGY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3196
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.