Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM4SF1/L6 Antibody (877621), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB310981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310981 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310981 Supplier Novus Biologicals Supplier No. FAB8164PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TM4SF1/L6 Monoclonal specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TM4SF1/L6
Applications Flow Cytometry
Classification Monoclonal
Clone 877621
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias L6, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TM4SF1/L6, Met1-Cys202, Accession # P30408
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4071
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.