Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM4SF18 Antibody (211116) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB372168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372168 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372168 Supplier Novus Biologicals Supplier No. FAB3509I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TM4SF18 Monoclonal specifically detects TM4SF18 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TM4SF18
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 211116
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias L6D
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TM4SF18, Met1-Ile201, Accession # Q96CE8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116441
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.