Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM4SF4 Antibody (832441) [PerCP], Novus Biologicals™
SDP

Catalog No. NB373762 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373762 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373762 Supplier Novus Biologicals Supplier No. FAB7998C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TM4SF4 Monoclonal specifically detects TM4SF4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TM4SF4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 832441
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FLJ31015, il-TMP, ILTMPintestinal and liver (il) tetraspan membrane protein, Intestine and liver tetraspan membrane protein, transmembrane 4 L six family member 4, transmembrane 4 L6 family member 4, transmembrane 4 superfamily member 4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TM4SF4, Accession # P48230
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7104
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.