Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM7SF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | TM7SF2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TM7SF2 Polyclonal specifically detects TM7SF2 in Human samples. It is validated for Western Blot.Specifications
| TM7SF2 | |
| Polyclonal | |
| Rabbit | |
| NP_003264 | |
| 7108 | |
| Synthetic peptide directed towards the N terminal of human TM7SF2. Peptide sequence LAARSGPARLLGPPASLPGLEVLWSPRALLLWLAWLGLQAALYLLPARKV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ANG1, C-14 sterol reductase, delta(14)-sterol reductase, delta-14-SR, DHCR14A, NET47, putative sterol reductase SR-1, sterol C14-reductase, transmembrane 7 superfamily member 2 | |
| TM7SF2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title