Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM7SF2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985988
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5988 Supplier Bioss Supplier No. BS4221R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TM7SF2 is involved in the conversion of lanosterol to cholesterol.

Specifications

Antigen TM7SF2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TM7SF2
Gene Accession No. O76062
Gene Alias 3110041O18Rik; 3-beta-hydroxysterol Delta (14)-reductase; ANG1; Another new gene 1 protein; BOS_24949; C-14 sterol reductase; C14SR; delta(14)-sterol reductase; Delta(14)-sterol reductase TM7SF2; Delta-14-SR; Dhcr14; DHCR14A; lamin B receptor-like; LOC100354153; NET47; Putative sterol reductase SR-1; Sterol C14-reductase; Tm7sf2; Transmembrane 7 superfamily member 2
Gene Symbols TM7SF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TM7SF2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7108
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.