Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMCO6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15766020UL
Description
TMCO6 Polyclonal specifically detects TMCO6 in Human samples. It is validated for Western Blot.Specifications
TMCO6 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q96DC7 | |
TMCO6 | |
Synthetic peptides corresponding to TMCO6(transmembrane and coiled-coil domains 6) The peptide sequence was selected from the N terminal of TMCO6. Peptide sequence LRQAQRGTEEKEREGALVSLRRGLQHPETQQTFIRLEGSMRTLVGLLTSN. | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ39769, PRO1580, transmembrane and coiled-coil domain-containing protein 6, transmembrane and coiled-coil domains 6 | |
Rabbit | |
Affinity Purified | |
RUO | |
55374 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction