Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMCO6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TMCO6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157660
![]() |
Novus Biologicals
NBP157660 |
100 μL |
Each for $487.50
|
|
|||||
NBP15766020
![]() |
Novus Biologicals
NBP15766020UL |
20 μL | N/A | N/A | N/A | ||||
Description
TMCO6 Polyclonal specifically detects TMCO6 in Human samples. It is validated for Western Blot.Specifications
TMCO6 | |
Polyclonal | |
Rabbit | |
Q96DC7 | |
55374 | |
Synthetic peptides corresponding to TMCO6(transmembrane and coiled-coil domains 6) The peptide sequence was selected from the N terminal of TMCO6. Peptide sequence LRQAQRGTEEKEREGALVSLRRGLQHPETQQTFIRLEGSMRTLVGLLTSN. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ39769, PRO1580, transmembrane and coiled-coil domain-containing protein 6, transmembrane and coiled-coil domains 6 | |
TMCO6 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title