Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMED1 Antibody (1009527), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313668 Supplier Novus Biologicals Supplier No. FAB2243APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMED1 Monoclonal specifically detects TMED1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TMED1
Applications Flow Cytometry
Classification Monoclonal
Clone 1009527
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias Il1rl1l, IL1RL1LGIL1RL1-binding protein, interleukin 1 receptor-like 1 ligand, Interleukin-1 receptor-like 1 ligand, MGC1270, Putative T1/ST2 receptor-binding protein, ST2L, T1/ST2 receptor binding protein, transmembrane emp24 domain containing 1, transmembrane emp24 domain-containing protein 1, transmembrane emp24 protein transport domain containing 1
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293 derived human TMED1, Met1-Asn194, Accession # Q13445
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11018
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.