Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567440 Supplier Invitrogen™ Supplier No. PA567440
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPQGNTIYRET KKQYDSFTYR AEVKGVYQF Highest antigen sequence identity to the following orthologs - mouse 80%, rat 83%.

Potential role in vesicular protein trafficking, mainly in the early secretory pathway. Contributes to the coupled localization of TMED2 and TMED10 in the cis-Golgi network. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMED3
Gene Accession No. Q9Y3Q3
Gene Alias 1200002G13Rik; AW546672; C15orf22; integral type I protein; membrane protein p24B; MGC133022; p24 family protein gamma-4; P24b; p24g4; p24gamma4; p26; testis tissue sperm-binding protein Li 48e; TMED3; transmembrane emp24 domain containing 3; transmembrane emp24 domain-containing protein 3; transmembrane emp24 domain-containing protein 3-like; transmembrane emp24 protein transport domain containing 3; transmembrane p24 trafficking protein 3; UNQ5357/PRO1078
Gene Symbols TMED3
Host Species Rabbit
Immunogen Recombinant Human TMED3. Recombinant protein control fragment (Product #RP-105602).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.