Learn More
Description
Specifications
Specifications
| Antigen | TMED4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml |
| Formulation | PBS containing 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | Q7Z7H5 |
| Gene Alias | Endoplasmic reticulum stress-response protein 25, endoplasmic reticulum stress-response protein 25 kDa, ERS25, HNLF, Putative NF-kappa-B-activating protein 156, putative NFkB activating protein HNLF, transmembrane emp24 domain-containing protein 4, transmembrane emp24 protein transport domain containing 4 |
| Gene Symbols | TMED4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
