Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEFF1/Tomoregulin-1 Antibody (MM0571-7S12), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP211926UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211926UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211926UV Supplier Novus Biologicals Supplier No. NBP211926UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMEFF1/Tomoregulin-1 Monoclonal specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TMEFF1/Tomoregulin-1
Applications Western Blot
Classification Monoclonal
Clone MM0571-7S12
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C9orf2, H7365member 1, tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1
Gene Symbols TMEFF1
Host Species Mouse
Immunogen Human recombinant TMEFF1/Tomoregulin-1
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8577
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.