Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEFF2/Tomoregulin-2 Antibody (6G6), Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP326283100UL
Description
TMEFF2/Tomoregulin-2 Monoclonal antibody specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for ELISASpecifications
TMEFF2/Tomoregulin-2 | |
Monoclonal | |
Unconjugated | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
Human | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
6G6 | |
ELISA | |
CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2 | |
Recombinant Human TMEFF2/Tomoregulin-2 protein [UniProt Q9UIK5] | |
100 μL | |
Cancer, Cell Biology, Cytokines and Growth Factors | |
23671 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction