Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEFF2/Tomoregulin-2 Antibody (6G6), Novus Biologicals™
SDP

Catalog No. p-200022785 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298410 50 μL
NB298411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298410 Supplier Novus Biologicals Supplier No. NBP32628350UL
Only null left
Add to Cart
Add to Cart

Human Monoclonal Antibody

TMEFF2/Tomoregulin-2 Monoclonal antibody specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for ELISA

Specifications

Antigen TMEFF2/Tomoregulin-2
Applications ELISA
Classification Monoclonal
Clone 6G6
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2
Host Species Human
Immunogen Recombinant Human TMEFF2/Tomoregulin-2 protein [UniProt Q9UIK5]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytokines and Growth Factors
Primary or Secondary Primary
Gene ID (Entrez) 23671
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.