Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEFF2/Tomoregulin-2 Antibody (SR1696), Novus Biologicals™
SDP

Catalog No. NB260199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260199 100 μg
NB259327 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260199 Supplier Novus Biologicals Supplier No. NBP322256100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TMEFF2/Tomoregulin-2 Monoclonal antibody specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen TMEFF2/Tomoregulin-2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1696
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEFF2/Tomoregulin-2 (Uniprot #: Q9UIK5)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytokines and Growth Factors
Primary or Secondary Primary
Gene ID (Entrez) 23671
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.