Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM107 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB192879 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB192879 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB192879 Supplier Novus Biologicals Supplier No. NBP281865F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM107 Polyclonal antibody specifically detects TMEM107 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TMEM107
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias transmembrane protein 107
Host Species Rabbit
Immunogen TMEM107 antibody was raised against a 19 amino acid peptide near the amino terminus of human TMEM107. The immunogen is located within amino acids 20 - 70 of TMEM107 . Amino Acid Squence: LPLTFTPEEYDKQDIHPL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 84314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.