Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM119 Antibody (1023426), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB340090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340090 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340090 Supplier Novus Biologicals Supplier No. FAB10313MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMEM119 Monoclonal specifically detects TMEM119 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen TMEM119
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 1023426
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias OBIF, Osteoblast Induction Factor, TMEM119
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO derived human TREM-119, Arg26-Met96, Accession # Q4V9L6.1
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338773
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.