Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM119 Antibody (2699A), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB305773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305773 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305773 Supplier Novus Biologicals Supplier No. FAB103131E
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry

Specifications

Antigen TMEM119
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 2699A
Conjugate DyLight 405
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias OBIF, Osteoblast Induction Factor, TMEM119
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO derived human TREM119, Arg26-Met96, Accession # Q4V9L6.1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338773
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.