Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM119 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TMEM119 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5707
![]() ![]() |
Proteintech Group Inc
275851AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
27585-1-AP recognizes a 45-50 kDa band likely due to glycosylation.
TMEM119 immunohistochemistry might provide a useful tool for investigating the biology and pathology of human microglia. The predicted MW of TMEM119 is 29 kDa.Specifications
TMEM119 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q4V9L6 | |
338773 | |
TMEM119 Fusion Protein Ag26269 | |
Primary | |
-20°C | |
TMEM119 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
OBIF, Osteoblast induction factor, TMEM119, transmembrane protein 119, UNQ731/PRO1415 | |
TMEM119 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title