Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM126B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | TMEM126B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15976820
![]() |
Novus Biologicals
NBP15976820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159768
![]() |
Novus Biologicals
NBP159768 |
100 μL |
Each for $487.50
|
|
|||||
Description
TMEM126B Polyclonal specifically detects TMEM126B in Human samples. It is validated for Western Blot.Specifications
TMEM126B | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
HT007, MGC111203, transmembrane protein 126B | |
TMEM126B | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q8IUX1 | |
55863 | |
Synthetic peptides corresponding to TMEM126B(transmembrane protein 126B) The peptide sequence was selected from the N terminal of TMEM126B. Peptide sequence AASMHGQPSPSLEDAKLRRPMVIEIIEKNFDYLRKEMTQNIYQMATFGTT. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title