Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM147 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17933420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17933420 20 μL
NBP179334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17933420 Supplier Novus Biologicals Supplier No. NBP17933420UL

Rabbit Polyclonal Antibody

TMEM147 Polyclonal specifically detects TMEM147 in Rat samples. It is validated for Western Blot.

Specifications

Antigen TMEM147
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_001033583
Gene Alias MGC1936, NIFIE14, Protein NIFIE 14, seven transmembrane domain protein, transmembrane protein 147
Gene Symbols TMEM147
Host Species Rabbit
Immunogen The immunogen for this antibody is Tmem147. Peptide sequence VTYLFVQLCKMLFLATFFPTWEGGIYDFIGEFMKASVDVADLIGLNLVMS.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10430
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.