Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM156 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB183026 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183026 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183026 Supplier Novus Biologicals Supplier No. NBP306638DL594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM156 Polyclonal antibody specifically detects TMEM156 in Human samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen TMEM156
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ23235, transmembrane protein 156
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human TMEM156 (AAH30803.1; Lys26-Ser211).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80008
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.