Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM168 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17072720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17072720 20 μL
NBP170727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17072720 Supplier Novus Biologicals Supplier No. NBP17072720UL

Rabbit Polyclonal Antibody

TMEM168 Polyclonal specifically detects TMEM168 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen TMEM168
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Alias FLJ13576, transmembrane protein 168
Gene Symbols TMEM168
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TMEM168(transmembrane protein 168) The peptide sequence was selected from the C terminal of TMEM168. Peptide sequence EEADPPQLGDFTKDWVEYNCNSSNNICWTEKGRTVKAVYGVSKRWSDYTL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64418
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.