Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM169 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB052755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052755 Supplier Novus Biologicals Supplier No. NBP298157F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM169 Polyclonal specifically detects TMEM169 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TMEM169
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias DKFZp781L2456, FLJ34263, transmembrane protein 169
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TMEM169 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92691
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.