Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM176A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | TMEM176A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TMEM176A Polyclonal specifically detects TMEM176A in Human samples. It is validated for Western Blot.Specifications
| TMEM176A | |
| Polyclonal | |
| Rabbit | |
| Q96HP8 | |
| 55365 | |
| Synthetic peptides corresponding to TMEM176A(transmembrane protein 176A) The peptide sequence was selected from the middle region of TMEM176A. Peptide sequence GYSYYNSACRISSSSDWNTPAPTQSPEEVRRLHLCTSFMDMLKALFRTLQ. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| GS188, HCA112Hepatocellular carcinoma-associated antigen 112, likley ortholog of mouse GS188, transmembrane protein 176A | |
| TMEM176A | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title