Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM184B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17919620UL
Description
TMEM184B Polyclonal specifically detects TMEM184B in Rat samples. It is validated for Western Blot.Specifications
TMEM184B | |
Polyclonal | |
Western Blot 1:1000 | |
NP_001166841 | |
TMEM184B | |
The immunogen for this antibody is Tmem184b. Peptide sequence STVILQAFGKYRDGDFDVTSGYLYVTIIYNISVSLALYALFLFYFATREL. | |
20 μL | |
Primary | |
Rat | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
DKFZp586A1024, DKFZp686F07174, DKFZp686N1150, FM08, PSEC0108, transmembrane protein 184B | |
Rabbit | |
Affinity Purified | |
RUO | |
25829 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction