Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM184B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | TMEM184B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17919620
![]() |
Novus Biologicals
NBP17919620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179196
![]() |
Novus Biologicals
NBP179196 |
100 μL |
Each for $487.50
|
|
|||||
Description
TMEM184B Polyclonal specifically detects TMEM184B in Rat samples. It is validated for Western Blot.Specifications
TMEM184B | |
Polyclonal | |
Rabbit | |
NP_001166841 | |
25829 | |
The immunogen for this antibody is Tmem184b. Peptide sequence STVILQAFGKYRDGDFDVTSGYLYVTIIYNISVSLALYALFLFYFATREL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp586A1024, DKFZp686F07174, DKFZp686N1150, FM08, PSEC0108, transmembrane protein 184B | |
TMEM184B | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title