Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM192 Antibody (21G8), Novus Biologicals™
SDP

Catalog No. NB298415 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298415 100 μL
NB298414 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298415 Supplier Novus Biologicals Supplier No. NBP326281100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TMEM192 Monoclonal antibody specifically detects TMEM192 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen TMEM192
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 21G8
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias FLJ38482, transmembrane protein 192
Host Species Rabbit
Immunogen A synthesized peptide derived from Human TMEM192 [UniProt Q8IY95]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201931
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.