Learn More
Description
Specifications
Specifications
| Antigen | TMEM214 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunoprecipitation 15-20 μL/mg lysate |
| Formulation | Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide |
| Gene Alias | FLJ20254, FLJ39682, PP446, transmembrane protein 214 |
| Gene Symbols | TMEM214 |
| Host Species | Rabbit |
| Immunogen | Immunogen sequence between amino acids 50 and 100. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
