Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM231 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB352674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352674 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352674 Supplier Novus Biologicals Supplier No. NBP306280APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM231 Polyclonal antibody specifically detects TMEM231 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TMEM231
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ALYE870, FLJ22167, PRO1886, transmembrane protein 231
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TMEM231 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79583
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.