Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM26 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 8 publications

Supplier:  Novus Biologicals NBP227334

 View more versions of this product

Catalog No. NBP227334


Only null left
Add to Cart

Description

Description

TMEM26 Polyclonal specifically detects TMEM26 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TMEM26
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
DKFZp686D09128, Em:AC068892.1, MGC35010, transmembrane protein 26
Rabbit
Protein A purified
RUO
219623
Human, Mouse
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1 - 2 ug/mL, Simple Western 1:20, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 30089732), Immunohistochemistry-Paraffin reported in scientific literature (PMID 28250021)
Q6ZUK4
TMEM26
A portion of amino acids 300-350 of human Tmem26 was used as the immunogen.
0.1 mg
Primary
The amino acid sequence used as immunogen is 100% homologous in human, 89% homologous in chimpanzee and 73% homologous in monkey.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only