Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM26 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227334SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227334SS 0.025 mg
NBP227334 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227334SS Supplier Novus Biologicals Supplier No. NBP227334SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

TMEM26 Polyclonal specifically detects TMEM26 in Human, Mouse samples. It is validated for Flow Cytometry, IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen TMEM26
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, Simple Western 1:20, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 30089732), Immunohistochemistry-Paraffin reported in scientific literature (PMID 28250021)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q6ZUK4
Gene Alias DKFZp686D09128, Em:AC068892.1, MGC35010, transmembrane protein 26
Gene Symbols TMEM26
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human Tmem26 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219623
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 89% homologous in chimpanzee and 73% homologous in monkey.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.