Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM27 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 240591AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-4223


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

TMEM27 (Transmembrane protein 27) is also named as collectrin and belongs to the TMEM27 family. It is a pancreatic beta-cell surface protein promoting the preservation of functional beta-cell mass and may be responsible for the sodium retention in salt-sensitive hypertension. This full length has two glycosylation sites.
Specifications

Specifications

TMEM27
Polyclonal
Unconjugated
TMEM27
Collectrin, NX 17, NX17, TMEM27, transmembrane protein 27
Rabbit
Antigen Affinity Chromatography
RUO
57393
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HBJ8
TMEM27
TMEM27 Fusion Protein Ag21257
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.